Assay Details
| Gene Symbol: Tm9sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502058.2 | XP_003502106.1 | 3 - 4 | 1225 | 55 | |
| XM_003502059.2 | XP_003502107.1 | 3 - 4 | 1154 | 55 | ||
| GenBank mRNA | JI879037.1 | - | 3 - 4 | 1127 | 55 | |
| JP057976.1 | - | 3 - 4 | 1127 | 55 | ||
More Information
Set Membership: |
|

Tm9sf1
Genomic Map