Assay Details
| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497577.2 | XP_003497625.1 | 13 - 14 | 1121 | 68 | |
| GenBank mRNA | JI890004.1 | - | 13 - 14 | 1121 | 68 | |
| JP062663.1 | - | 13 - 14 | 1121 | 68 | ||
More Information
Set Membership: |
|

Asah1
Genomic Map