Assay Details
| Gene Symbol: LOC100765128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500361.2 | XP_003500409.1 | 3 - 4 | 316 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100765128| Gene Symbol: LOC100765128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500361.2 | XP_003500409.1 | 3 - 4 | 316 | 53 | |
Set Membership: |
|