Assay Details
| Gene Symbol: Osr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502107.2 | XP_003502155.1 | 2 - 3 | 977 | 103 | |
| XM_007642707.1 | XP_007640897.1 | 2 - 3 | 772 | 103 | ||
More Information
Set Membership: |
|

Osr1
Genomic Map