Assay Details
| Gene Symbol: Cd276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502396.2 | XP_003502444.2 | 7 - 8 | 1035 | 64 | |
| XM_007643020.1 | XP_007641210.1 | 7 - 8 | 1046 | 64 | ||
More Information
Set Membership: |
|

Cd276
Genomic Map