Assay Details
| Gene Symbol: Tgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509411.2 | XP_003509459.1 | 11 - 12 | 2459 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgs1| Gene Symbol: Tgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509411.2 | XP_003509459.1 | 11 - 12 | 2459 | 74 | |
Set Membership: |
|