Assay Details
| Gene Symbol: Pagr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510120.2 | XP_003510168.1 | 2 - 3 | 899 | 70 | |
| GenBank mRNA | JI870288.1 | - | 2 - 3 | 968 | 70 | |
| JP040170.1 | - | 2 - 3 | 968 | 70 | ||
More Information
Set Membership: |
|

Pagr1
Genomic Map