Assay Details
| Gene Symbol: Dpcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510344.2 | XP_003510392.1 | 4 - 5 | 449 | 71 | |
| GenBank mRNA | JI878588.1 | - | 4 - 5 | 436 | 71 | |
| JP062451.1 | - | 4 - 5 | 436 | 71 | ||
More Information
Set Membership: |
|

Dpcd
Genomic Map