Assay Details
| Gene Symbol: Gsk3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649494.1 | XP_007647684.1 | 1217 | 62 | ||
| GenBank mRNA | JI866681.1 | - | 1022 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsk3a| Gene Symbol: Gsk3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649494.1 | XP_007647684.1 | 1217 | 62 | ||
| GenBank mRNA | JI866681.1 | - | 1022 | 62 | ||
Set Membership: |
|