Assay Details
| Gene Symbol: Btf3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496500.2 | XP_003496548.2 | 2 - 3 | 133 | 81 | |
| XM_007650721.1 | XP_007648911.1 | 2 - 3 | 87 | 81 | ||
| GenBank mRNA | JI882079.1 | - | 1 - 2 | 74 | 81 | |
| JP044355.1 | - | 1 - 2 | 74 | 81 | ||
More Information
Set Membership: |
|

Btf3l4
Genomic Map