Assay Details
| Gene Symbol: Prkar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500312.2 | XP_003500360.1 | 3 - 4 | 466 | 76 | |
| GenBank mRNA | JI889185.1 | - | 4 - 5 | 556 | 76 | |
| JP040345.1 | - | 4 - 5 | 556 | 76 | ||
More Information
Set Membership: |
|

Prkar1a
Genomic Map