Assay Details
| Gene Symbol: Clu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505153.2 | XP_003505201.2 | 5 - 6 | 870 | 56 | |
| XM_007645697.1 | XP_007643887.1 | 5 - 6 | 925 | 56 | ||
| XM_007645698.1 | XP_007643888.1 | 5 - 6 | 967 | 56 | ||
| GenBank mRNA | JI889982.1 | - | 5 - 6 | 888 | 56 | |
More Information
Set Membership: |
|

Clu
Genomic Map