Assay Details
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510379.1 | XP_003510427.1 | 1 - 2 | 102 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtl5| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510379.1 | XP_003510427.1 | 1 - 2 | 102 | 84 | |