Assay Details
| Gene Symbol: Htra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511129.2 | XP_003511177.1 | 1 - 2 | 647 | 57 | |
| GenBank mRNA | JI886612.1 | - | 5 - 6 | 57 | ||
| JP054219.1 | - | 1 - 2 | 127 | 57 | ||
More Information
Set Membership: |
|

Htra2
Genomic Map