Assay Details
| Gene Symbol: LOC100760823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512730.1 | XP_003512778.1 | 1 | 1021 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100760823| Gene Symbol: LOC100760823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512730.1 | XP_003512778.1 | 1 | 1021 | 100 | |
Set Membership: |
|