Assay Details
| Gene Symbol: LOC100767645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607769.1 | XP_007605959.1 | 4 - 5 | 492 | 89 | |
| GenBank mRNA | JI877416.1 | - | 1 - 2 | 74 | 89 | |
| JP056586.1 | - | 1 - 2 | 74 | 89 | ||
More Information
Set Membership: |
|

LOC100767645
Genomic Map