Assay Details
| Gene Symbol: LOC100767645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607769.1 | XP_007605959.1 | 5 - 6 | 864 | 93 | |
| GenBank mRNA | JI877416.1 | - | 2 - 3 | 446 | 93 | |
| JP056586.1 | - | 2 - 3 | 446 | 93 | ||
More Information
Set Membership: |
|

LOC100767645
Genomic Map