Assay Details
| Gene Symbol: LOC100767645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607769.1 | XP_007605959.1 | 7 - 8 | 1101 | 68 | |
| GenBank mRNA | JI877416.1 | - | 4 - 5 | 683 | 68 | |
| JP056586.1 | - | 4 - 5 | 683 | 68 | ||
More Information
Set Membership: |
|

LOC100767645
Genomic Map