Assay Details
| Gene Symbol: LOC100767645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607769.1 | XP_007605959.1 | 9 - 10 | 1349 | 93 | |
| GenBank mRNA | JI877416.1 | - | 6 - 7 | 931 | 93 | |
| JP056586.1 | - | 6 - 7 | 931 | 93 | ||
More Information
Set Membership: |
|

LOC100767645
Genomic Map