Assay Details
| Gene Symbol: LOC100769195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643082.1 | XP_007641272.1 | 11 - 12 | 1148 | 85 | |
| GenBank mRNA | JP045526.1 | - | 10 - 11 | 1141 | 85 | |
More Information
Set Membership: |
|

LOC100769195
Genomic Map