Assay Details
| Gene Symbol: LOC100769195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643082.1 | XP_007641272.1 | 16 - 17 | 2005 | 67 | |
| GenBank mRNA | JP045526.1 | - | 15 - 16 | 1998 | 67 | |
More Information
Set Membership: |
|

LOC100769195
Genomic Map