Assay Details
| Gene Symbol: LOC100769195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643082.1 | XP_007641272.1 | 22 - 23 | 2706 | 66 | |
| GenBank mRNA | JP045526.1 | - | 21 - 22 | 2699 | 66 | |
More Information
Set Membership: |
|

LOC100769195
Genomic Map