Assay Details
| Gene Symbol: Uck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652783.1 | XP_007650973.1 | 3 - 4 | 399 | 72 | |
| GenBank mRNA | JP062088.1 | - | 1 - 2 | 100 | 72 | |
More Information
Set Membership: |
|

Uck1
Genomic Map