Assay Details
| Gene Symbol: LOC103161503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644166.1 | XP_007642356.1 | 4 - 5 | 266 | 80 | |
| GenBank mRNA | JI884694.1 | - | 4 - 5 | 268 | 80 | |
More Information
Set Membership: |
|

LOC103161503
Genomic Map