Assay Details
| Gene Symbol: LOC103161503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644166.1 | XP_007642356.1 | 9 - 10 | 793 | 70 | |
| GenBank mRNA | JI884694.1 | - | 9 - 10 | 795 | 70 | |
More Information
Set Membership: |
|

LOC103161503
Genomic Map