Assay Details
| Gene Symbol: Sage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640578.1 | XP_007638768.1 | 8 - 9 | 1140 | 128 | |
| GenBank mRNA | JP044506.1 | - | 1 - 2 | 111 | 128 | |
More Information
Set Membership: |
|

Sage1
Genomic Map