Assay Details
| Gene Symbol: Sage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640578.1 | XP_007638768.1 | 10 - 11 | 1488 | 100 | |
| GenBank mRNA | JI893422.1 | - | 3 - 4 | 375 | 100 | |
| JP044506.1 | - | 3 - 4 | 459 | 100 | ||
More Information
Set Membership: |
|

Sage1
Genomic Map