Assay Details
| Gene Symbol: Sage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640578.1 | XP_007638768.1 | 13 - 14 | 2231 | 104 | |
| GenBank mRNA | JI893031.1 | - | 1 - 2 | 205 | 104 | |
| JP066081.1 | - | 1 - 2 | 205 | 104 | ||
More Information
Set Membership: |
|

Sage1
Genomic Map