Assay Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649010.1 | XP_007647200.1 | 10 - 11 | 1623 | 68 | |
| XM_007649011.1 | XP_007647201.1 | 10 - 11 | 1623 | 68 | ||
| GenBank mRNA | JP059895.1 | - | 10 - 11 | 1639 | 68 | |
More Information
Set Membership: |
|

Hif1a
Genomic Map