Assay Details
| Gene Symbol: LOC100766027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647266.1 | XP_007645456.1 | 10 - 11 | 1184 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100766027| Gene Symbol: LOC100766027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647266.1 | XP_007645456.1 | 10 - 11 | 1184 | 98 | |
Set Membership: |
|