Assay Details
| Gene Symbol: LOC100755423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653603.1 | XP_007651793.1 | 5 - 6 | 1586 | 102 | |
| XM_007653604.1 | XP_007651794.1 | 4 - 5 | 1967 | 102 | ||
| XM_007653605.1 | XP_007651795.1 | 5 - 6 | 1475 | 102 | ||
| GenBank mRNA | JI893682.1 | - | 2 - 3 | 307 | 102 | |
| JP063791.1 | - | 3 - 4 | 442 | 102 | ||
More Information
Set Membership: |
|

LOC100755423
Genomic Map