Assay Details
| Gene Symbol: LOC100755423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653603.1 | XP_007651793.1 | 6 - 7 | 2238 | 65 | |
| XM_007653604.1 | XP_007651794.1 | 5 - 6 | 2619 | 65 | ||
| XM_007653605.1 | XP_007651795.1 | 6 - 7 | 2127 | 65 | ||
| GenBank mRNA | JI893682.1 | - | 3 - 4 | 959 | 65 | |
| JP063791.1 | - | 4 - 5 | 1094 | 65 | ||
More Information
Set Membership: |
|

LOC100755423
Genomic Map