Assay Details
| Gene Symbol: Nsmce4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513285.2 | XP_003513333.1 | 1 - 2 | 382 | 62 | |
| XM_007653592.1 | XP_007651782.1 | 1 - 2 | 382 | 62 | ||
| GenBank mRNA | JI869428.1 | - | 1 - 2 | 93 | 62 | |
More Information
Set Membership: |
|

Nsmce4a
Genomic Map