Assay Details
| Gene Symbol: LOC100767045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653228.1 | XP_007651418.1 | 14 - 15 | 2220 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100767045| Gene Symbol: LOC100767045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653228.1 | XP_007651418.1 | 14 - 15 | 2220 | 109 | |
Set Membership: |
|