Assay Details
| Gene Symbol: LOC100767045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653228.1 | XP_007651418.1 | 27 - 28 | 4653 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100767045| Gene Symbol: LOC100767045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653228.1 | XP_007651418.1 | 27 - 28 | 4653 | 81 | |
Set Membership: |
|