Assay Details
| Gene Symbol: Cenpv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649487.1 | XP_007647677.1 | 5 - 6 | 642 | 73 | |
| GenBank mRNA | JI882997.1 | - | 3 - 4 | 489 | 73 | |
More Information
Set Membership: |
|

Cenpv
Genomic Map