Assay Details
Gene Symbol: Lipe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649490.1 | XP_007647680.1 | 5 - 6 | 549 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
