Assay Details
Gene Symbol: Nrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496017.2 | XP_003496065.1 | 2 - 3 | 480 | 67 | |
XM_007646261.1 | XP_007644451.1 | 2 - 3 | 480 | 67 | ||
XM_007646270.1 | XP_007644460.1 | 2 - 3 | 402 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
