Assay Details
Gene Symbol: Sharpin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507672.2 | XP_003507720.1 | 5 - 6 | 1190 | 69 | |
XM_007648245.1 | XP_007646435.1 | 5 - 6 | 1195 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
