Assay Details
| Gene Symbol: Kctd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510231.2 | XP_003510279.2 | 1 - 2 | 291 | 73 | |
| XM_007650845.1 | XP_007649035.1 | 2 - 3 | 1663 | 73 | ||
| XM_007650846.1 | XP_007649036.1 | 1 - 2 | 86 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007650845.1, XM_007650846.1 transcripts.
Set Membership: |
|

Kctd6
Genomic Map