Assay Details
| Gene Symbol: Cenpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512341.2 | XP_003512389.1 | 1 - 2 | 148 | 66 | |
| XM_003512342.2 | XP_003512390.1 | 1 - 2 | 182 | 66 | ||
More Information
Set Membership: |
|

Cenpm
Genomic Map