Assay Details
| Gene Symbol: Aldh3b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650570.1 | XP_007648760.1 | 6 - 7 | 930 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh3b2| Gene Symbol: Aldh3b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650570.1 | XP_007648760.1 | 6 - 7 | 930 | 62 | |
Set Membership: |
|