Assay Details
| Gene Symbol: LOC100768019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654769.1 | XP_007652959.1 | 2 - 3 | 182 | 108 | |
| XM_007654770.1 | XP_007652960.1 | 2 - 3 | 185 | 108 | ||
More Information
Set Membership: |
|

LOC100768019
Genomic Map