Assay Details
Gene Symbol: Il12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502207.1 | XP_003502255.1 | 5 - 6 | 854 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
