Assay Details
| Gene Symbol: Rnase4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509210.2 | XP_003509258.2 | 3 - 4 | 461 | 77 | |
| XM_007649803.1 | XP_007647993.1 | 3 - 4 | 461 | 77 | ||
| XM_007649804.1 | XP_007647994.1 | 2 - 3 | 360 | 77 | ||
| GenBank mRNA | JI865387.1 | - | 1 - 2 | 338 | 77 | |
More Information
Set Membership: |
|

Rnase4
Genomic Map