Assay Details
| Gene Symbol: March2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510501.2 | XP_003510549.1 | 3 - 4 | 553 | 96 | |
| XM_007651130.1 | XP_007649320.1 | 3 - 4 | 554 | 96 | ||
| XM_007651131.1 | XP_007649321.1 | 3 - 4 | 576 | 96 | ||
| GenBank mRNA | JI861988.1 | - | 2 - 3 | 296 | 96 | |
| JP050292.1 | - | 2 - 3 | 296 | 96 | ||
More Information
Set Membership: |
|

March2
Genomic Map