Assay Details
| Gene Symbol: LOC100752257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512014.1 | XP_003512062.1 | 5 - 6 | 745 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100752257| Gene Symbol: LOC100752257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512014.1 | XP_003512062.1 | 5 - 6 | 745 | 88 | |
Set Membership: |
|