Assay Details
| Gene Symbol: LOC100762081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512228.2 | XP_003512276.2 | 4 - 5 | 251 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100762081| Gene Symbol: LOC100762081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512228.2 | XP_003512276.2 | 4 - 5 | 251 | 107 | |
Set Membership: |
|