Assay Details
| Gene Symbol: LOC100755731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JP045289.1 | - | 1176 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100755731| Gene Symbol: LOC100755731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JP045289.1 | - | 1176 | 70 | ||