Assay Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648386.1 | XP_007646576.1 | 3 - 4 | 726 | 88 | |
| XM_007648387.1 | XP_007646577.1 | 3 - 4 | 672 | 88 | ||
| XM_007648388.1 | XP_007646578.1 | 3 - 4 | 660 | 88 | ||
| XM_007648389.1 | XP_007646579.1 | 3 - 4 | 659 | 88 | ||
| XM_007648390.1 | XP_007646580.1 | 3 - 4 | 726 | 88 | ||
| XM_007648391.1 | XP_007646581.1 | 3 - 4 | 726 | 88 | ||
| GenBank mRNA | JI864007.1 | - | 2 - 3 | 699 | 88 | |

Pvrl3
Genomic Map